siRNA Details
| Gene Symbol: LOC654780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095331.1 | - | 3 | 1008 | ||
| AK123582.1 | - | 5 | 870 | |||
| DQ323994.1 | - | 2 | 390 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC654780| Gene Symbol: LOC654780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095331.1 | - | 3 | 1008 | ||
| AK123582.1 | - | 5 | 870 | |||
| DQ323994.1 | - | 2 | 390 | |||