siRNA Details
Gene Symbol: LOC105377267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136191.1 | 6 | 2808 | |||
GenBank mRNA | AK124272.1 | - | 4 | 2483 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505646.1 | - | 4 | 2483 | Chr. 4 - 69200420 - 69216766 [+] on Build GRCh38 |
