siRNA Details
Gene Symbol: LINC01262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121679.1 | 3 | 658 | |||
GenBank mRNA | BC092460.1 | - | 3 | 658 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000513681.1 | - | 3 | 659 | Chr. 4 - 189659605 - 189661486 [+] on Build GRCh38 |
