siRNA Details
| Gene Symbol: LOC100128253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056228.1 | - | 2 | 1699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100128253| Gene Symbol: LOC100128253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056228.1 | - | 2 | 1699 | ||