siRNA Details
| Gene Symbol: LINC01365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110660.1 | 1 | 25 | |||
| GenBank mRNA | BC038750.1 | - | 1 | 25 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000503266.5 | - | 1 | 25 | Chr. 4 - 119799089 - 119804515 [-] on Build GRCh38 | |

LINC01365
Genomic Map