siRNA Details
| Gene Symbol: LOC102724596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110883.1 | 5 | 502 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000511322.1 | - | 2 | 189 | Chr. 17 - 49375380 - 49380094 [+] on Build GRCh38 | |

LOC102724596
Genomic Map