siRNA Details
Gene Symbol: TMEM108-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110812.1 | 3 | 1415 | |||
GenBank mRNA | BC037929.2 | - | 3 | 1416 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505721.1 | - | 3 | 1482 | Chr. 3 - 133246426 - 133257188 [-] on Build GRCh38 |
