siRNA Details
| Gene Symbol: WWC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126473.1 | 2 | 270 | |||
| GenBank mRNA | BX537950.1 | - | 2 | 271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WWC2-AS1| Gene Symbol: WWC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126473.1 | 2 | 270 | |||
| GenBank mRNA | BX537950.1 | - | 2 | 271 | ||