siRNA Details
| Gene Symbol: LOC101927934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105045.1 | 2 | 745 | |||
| GenBank mRNA | BC032795.1 | - | 2 | 694 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000515627.2 | - | 2 | 745 | Chr. 5 - 134506552 - 134509229 [+] on Build GRCh38 | |

LOC101927934
Genomic Map