siRNA Details
| Gene Symbol: LOC101929019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125910.1 | 1 | 110 | |||
| GenBank mRNA | BC042433.1 | - | 1 | 81 | ||
| BQ441613.1 | - | 113 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929019| Gene Symbol: LOC101929019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125910.1 | 1 | 110 | |||
| GenBank mRNA | BC042433.1 | - | 1 | 81 | ||
| BQ441613.1 | - | 113 | ||||