siRNA Details
| Gene Symbol: LOC101929019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125910.1 | 4 | 361 | |||
| GenBank mRNA | BC042433.1 | - | 4 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929019| Gene Symbol: LOC101929019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125910.1 | 4 | 361 | |||
| GenBank mRNA | BC042433.1 | - | 4 | 332 | ||