siRNA Details
| Gene Symbol: LOC101927845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125427.1 | 1 | 651 | |||
| GenBank mRNA | AK057402.1 | - | 1 | 651 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505083.1 | - | 1 | 651 | Chr. 8 - 134849935 - 134881899 [+] on Build GRCh38 | |

LOC101927845
Genomic Map