siRNA Details
| Gene Symbol: LVCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131958.1 | 2 | 576 | |||
| NR_131959.1 | 2 | 469 | ||||
| GenBank mRNA | AL038572.2 | - | 2 | 468 | ||
| BC042992.1 | - | 2 | 581 | |||
| BU569738.1 | - | 2 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LVCAT1| Gene Symbol: LVCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131958.1 | 2 | 576 | |||
| NR_131959.1 | 2 | 469 | ||||
| GenBank mRNA | AL038572.2 | - | 2 | 468 | ||
| BC042992.1 | - | 2 | 581 | |||
| BU569738.1 | - | 2 | 388 | |||