siRNA Details
| Gene Symbol: LINC00958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038904.1 | 2 | 213 | |||
| GenBank mRNA | AK001432.1 | - | 2 | 213 | ||
| BQ929741.1 | - | 3 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00958| Gene Symbol: LINC00958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038904.1 | 2 | 213 | |||
| GenBank mRNA | AK001432.1 | - | 2 | 213 | ||
| BQ929741.1 | - | 3 | 259 | |||