siRNA Details
Gene Symbol: LINC00958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038904.1 | 2 | 1448 | |||
GenBank mRNA | AK001432.1 | - | 2 | 1447 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000504230.2 | - | 2 | 1448 | Chr. 11 - 12979535 - 12989548 [-] on Build GRCh38 |
