siRNA Details
| Gene Symbol: LINC01301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103854.1 | 3 | 2114 | |||
| GenBank mRNA | AK124262.1 | - | 3 | 2115 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000506428.1 | - | 3 | 2114 | Chr. 8 - 60403212 - 60413818 [-] on Build GRCh38 | |

LINC01301
Genomic Map