siRNA Details
Gene Symbol: LINC01256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126401.1 | 2 | 269 | |||
GenBank mRNA | BX088800.1 | - | 2 | 269 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000513270.1 | - | 2 | 269 | Chr. 4 - 132591089 - 132678503 [+] on Build GRCh38 |
