siRNA Details
| Gene Symbol: LINC01256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126401.1 | 4 | 446 | |||
| GenBank mRNA | BX088800.1 | - | 4 | 446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000513270.1 | - | 4 | 446 | Chr. 4 - 132591089 - 132678503 [+] on Build GRCh38 | |

LINC01256
Genomic Map