siRNA Details
Gene Symbol: LOC100507487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125882.1 | 5 | 843 | |||
GenBank mRNA | AK093416.1 | - | 5 | 789 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000511497.5 | - | 5 | 789 | Chr. 4 - 128428070 - 128519394 [+] on Build GRCh38 |
