siRNA Details
| Gene Symbol: LOC101927379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109881.1 | 8 | 836 | |||
| GenBank mRNA | BC043373.1 | - | 8 | 835 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000511194.5 | - | 8 | 835 | Chr. 5 - 122628952 - 122730559 [-] on Build GRCh38 | |

LOC101927379
Genomic Map