siRNA Details
| Gene Symbol: LOC101927059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134282.1 | 2 | 256 | |||
| GenBank mRNA | BC038569.1 | - | 2 | 256 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000506392.1 | - | 2 | 256 | Chr. 5 - 114576041 - 114579970 [+] on Build GRCh38 | |

LOC101927059
Genomic Map