siRNA Details
Gene Symbol: LOC101927059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134282.1 | 2 | 99 | |||
GenBank mRNA | BC038569.1 | - | 2 | 99 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000506392.1 | - | 2 | 99 | Chr. 5 - 114576041 - 114579970 [+] on Build GRCh38 |
