siRNA Details
| Gene Symbol: LOC100287592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104653.1 | 1 | 124 | |||
| GenBank mRNA | BM726851.1 | - | 1 | 144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100287592| Gene Symbol: LOC100287592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104653.1 | 1 | 124 | |||
| GenBank mRNA | BM726851.1 | - | 1 | 144 | ||