siRNA Details
| Gene Symbol: LOC105374366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134664.1 | 1 | 123 | |||
| NR_134665.1 | 1 | 123 | ||||
| GenBank mRNA | BX100005.1 | - | 1 | 105 | ||
| HY022020.1 | - | 1 | 121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374366| Gene Symbol: LOC105374366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134664.1 | 1 | 123 | |||
| NR_134665.1 | 1 | 123 | ||||
| GenBank mRNA | BX100005.1 | - | 1 | 105 | ||
| HY022020.1 | - | 1 | 121 | |||