siRNA Details
| Gene Symbol: LOC101927636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125886.1 | 7 | 735 | |||
| GenBank mRNA | BC029578.1 | - | 7 | 737 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927636| Gene Symbol: LOC101927636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125886.1 | 7 | 735 | |||
| GenBank mRNA | BC029578.1 | - | 7 | 737 | ||