siRNA Details
| Gene Symbol: LOC100507437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125384.1 | 3 | 1434 | |||
| NR_125385.1 | 2 | 1299 | ||||
| GenBank mRNA | AK123017.1 | - | 2 | 1299 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000504332.1 | - | 2 | 1299 | Chr. 14 - 105417581 - 105419739 [-] on Build GRCh38 | |

LOC100507437
Genomic Map