siRNA Details
| Gene Symbol: LOC105379178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086294.1 | - | 2 | 364 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000507389.1 | - | 2 | 359 | Chr. 5 - 133111055 - 133114475 [-] on Build GRCh38 | |

LOC105379178
Genomic Map