siRNA Details
Gene Symbol: LOC105379178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086294.1 | - | 2 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000507389.1 | - | 2 | 494 | Chr. 5 - 133111055 - 133114475 [-] on Build GRCh38 |
