siRNA Details
| Gene Symbol: LINC01609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125418.1 | 1 | 80 | |||
| GenBank mRNA | BI459905.1 | - | 1 | 85 | ||
| DB448904.1 | - | 1 | 48 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000522778.1 | - | 1 | 80 | Chr. 8 - 111183292 - 111236203 [-] on Build GRCh38 | |

LINC01609
Genomic Map