siRNA Details
Gene Symbol: LOC101929622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125820.1 | 2, 3 | 558 | |||
GenBank mRNA | BC031939.1 | - | 2, 3 | 558 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000523507.1 | - | 2, 3 | 558 | Chr. 8 - 37717579 - 37733756 [+] on Build GRCh38 |
