siRNA Details
| Gene Symbol: LINC00599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024281.1 | 3 | 2884 | |||
| GenBank mRNA | AF052108.1 | - | 1 | 1648 | ||
| AK055056.1 | - | 1 | 2144 | |||
| AK091593.1 | - | 1 | 2977 | |||
| AL832535.1 | - | 3 | 1609 | |||
| BC042097.1 | - | 2 | 831 | |||

LINC00599
Genomic Map