siRNA Details
| Gene Symbol: LINC00599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024281.1 | 1 | 1219 | |||
| GenBank mRNA | DA155571.1 | - | 1 | 480 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519461.1 | - | 1 | 1219 | Chr. 8 - 9900064 - 9903329 [-] on Build GRCh38 | |

LINC00599
Genomic Map