siRNA Details
Gene Symbol: DLGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103863.1 | 3 | 286 | |||
GenBank mRNA | AK093412.1 | - | 3 | 1546 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000518009.1 | - | 3 | 1546 | Chr. 8 - 1604552 - 1622417 [-] on Build GRCh38 |
