siRNA Details
| Gene Symbol: DLGAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093412.1 | - | 1 | 351 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000518009.1 | - | 1 | 351 | Chr. 8 - 1604552 - 1622417 [-] on Build GRCh38 | |

DLGAP2-AS1
Genomic Map