siRNA Details
| Gene Symbol: LINC01606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038235.2 | 10 | 1936 | |||
| GenBank mRNA | CR749704.1 | - | 8 | 1733 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519160.5 | - | 8 | 1733 | Chr. 8 - 57219089 - 57232636 [+] on Build GRCh38 | |

LINC01606
Genomic Map