siRNA Details
Gene Symbol: LINC01606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038235.2 | 10 | 1866 | |||
GenBank mRNA | CR749704.1 | - | 8 | 1663 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000519160.5 | - | 8 | 1663 | Chr. 8 - 57219089 - 57232636 [+] on Build GRCh38 |
