siRNA Details
| Gene Symbol: LOC101927766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109891.1 | 1 | 3360 | |||
| GenBank mRNA | AL833484.1 | - | 1 | 3360 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000522627.1 | - | 1 | 3360 | Chr. 5 - 159776772 - 159871384 [-] on Build GRCh38 | |

LOC101927766
Genomic Map