siRNA Details
| Gene Symbol: LOC105379194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134244.1 | 2 | 534 | |||
| NR_134245.1 | 2 | 390 | ||||
| GenBank mRNA | AI792600.1 | - | 2 | 390 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000520838.1 | - | 2 | 390 | Chr. 5 - 138744434 - 138753309 [-] on Build GRCh38 | |

LOC105379194
Genomic Map