siRNA Details
| Gene Symbol: LOC101927908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134280.1 | 2 | 458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927908| Gene Symbol: LOC101927908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134280.1 | 2 | 458 | |||