siRNA Details
Gene Symbol: CA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121632.1 | 1 | 53 | |||
NR_121633.1 | 1 | 53 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000524052.1 | - | 1 | 53 | Chr. 8 - 85456623 - 85463054 [-] on Build GRCh38 |
