siRNA Details
Gene Symbol: LOC101929341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134310.1 | 2 | 764 | |||
GenBank mRNA | BC065517.1 | - | 2 | 764 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000522541.1 | - | 2 | 764 | Chr. 8 - 52194362 - 52199580 [+] on Build GRCh38 |
