siRNA Details
Gene Symbol: LOC101929268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105002.1 | 4 | 1064 | |||
GenBank mRNA | AK092055.1 | - | 4 | 1064 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000522575.1 | - | 4 | 1064 | Chr. 8 - 48551567 - 48698510 [+] on Build GRCh38 |
