siRNA Details
| Gene Symbol: LOC101929268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105002.1 | 4 | 790 | |||
| GenBank mRNA | AK092055.1 | - | 4 | 790 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000522575.1 | - | 4 | 790 | Chr. 8 - 48551567 - 48698510 [+] on Build GRCh38 | |

LOC101929268
Genomic Map