siRNA Details
| Gene Symbol: LOC101929315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125809.1 | 3 | 290 | |||
| GenBank mRNA | BX096372.1 | - | 3 | 250 | ||
| HY022654.1 | - | 2 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929315| Gene Symbol: LOC101929315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125809.1 | 3 | 290 | |||
| GenBank mRNA | BX096372.1 | - | 3 | 250 | ||
| HY022654.1 | - | 2 | 412 | |||