siRNA Details
Gene Symbol: LOC101929315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125809.1 | 3 | 320 | |||
GenBank mRNA | BX096372.1 | - | 3 | 280 | ||
HY022654.1 | - | 2 | 442 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125809.1 | 3 | 320 | |||
GenBank mRNA | BX096372.1 | - | 3 | 280 | ||
HY022654.1 | - | 2 | 442 |