siRNA Details
| Gene Symbol: CTD-2270F17.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109899.1 | 3 | 1074 | |||
| GenBank mRNA | AK311026.1 | - | 3 | 1074 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000518387.1 | - | 3 | 1074 | Chr. 5 - 170389493 - 170422844 [+] on Build GRCh38 | |

CTD-2270F17.1
Genomic Map