siRNA Details
Gene Symbol: CTD-2270F17.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109899.1 | 3 | 534 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000518387.1 | - | 3 | 534 | Chr. 5 - 170389493 - 170422844 [+] on Build GRCh38 |
