siRNA Details
| Gene Symbol: LINC01592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039986.1 | 2 | 228 | |||
| GenBank mRNA | AK055514.1 | - | 2 | 228 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000518540.5 | - | 2 | 228 | Chr. 8 - 68911803 - 69104190 [-] on Build GRCh38 | |

LINC01592
Genomic Map