siRNA Details
Gene Symbol: LINC01484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074818.1 | - | 2 | 1881 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000521128.1 | - | 2 | 1891 | Chr. 5 - 173710440 - 173719692 [-] on Build GRCh38 |
