siRNA Details
| Gene Symbol: LOC101926908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110656.1 | 3 | 264 | |||
| GenBank mRNA | AK055677.1 | - | 3 | 264 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519427.1 | - | 3 | 264 | Chr. 8 - 72732045 - 72751843 [+] on Build GRCh38 | |

LOC101926908
Genomic Map